2-Iodoethanol stabilised for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.41353.9999
8.41353.9999EA 0 EUR
8.41353.9999
2-Iodoethanol stabilised for synthesis, Sigma-Aldrich®
2-Iodoethanol
Formula: C₂H₅IO
Boiling Pt: 85 °C (1013 hPa)
Density: 2,202 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00002830
CAS Number: 624-76-0
EINECS: 210-861-6
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Molar Mass 171.97 g/mol
Boiling Point 172 - 175 °C (1013 hPa)
Flash Point 65 °C
Density 2.202 g/cm3 (20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR