2-(4-Toluoyl)benzoic acid, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: 2-(4-Methylbenzoyl)benzenecarboxylic acid

M-Clarity™ quality level = MQ200

8.42258.0025
8.42258.0025EA 62.9 EUR
8.42258.0025
2-(4-Toluoyl)benzoic acid, Sigma-Aldrich®
2-(4-Toluoyl)benzoic acid
Formula: C₁₅H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00020287
CAS Number: 85-55-2
EINECS: 201-614-3

Order Now

Specification Test Results

Melting Point 133 - 137 °C
Molar Mass 240.26 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR