Gallic acid for synthesis, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3,4,5-Trihydroxybenzoic acid

M-Clarity™ quality level = MQ200

8.42649.0025 8.42649.0250 8.42649.9999
8.42649.0025EA 42.9 EUR
8.42649.0025 8.42649.0250 8.42649.9999
Gallic acid for synthesis, Sigma-Aldrich®
Gallic acid
Formula: C₇H₆O₅
MW: 170.12 g/mol
Boiling Pt: 501
Melting Pt: 251…252 °C
Density: 1.694 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002510
CAS Number: 149-91-7
EINECS: 205-749-9
Merck Index: 13,04366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR