DL-Tartaric acid for synthesis, Sigma-Aldrich®

Supplier: Merck
Warning


M-Clarity™ quality level = MQ200

8.43377.0100 8.43377.0500
8.43377.0100EA 35.8 EUR
8.43377.0100 8.43377.0500
DL-Tartaric acid for synthesis, Sigma-Aldrich®
DL-Tartaric acid
Formula: C₄H₆O₆
Storage Temperature: Ambient
MDL Number: MFCD00151254
CAS Number: 133-37-9
EINECS: 205-105-7

Order Now

Specification Test Results

Solubility 215 g/l (25 °C)
Melting Point 198...204 °C
Molar Mass 150.09 g/mol
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 210 °C
pH 1.6 (100 g/l, H2O, 25 °C)
Ignition Point 425 °C
pH value 1,6 (100 g/l, H2O, 25 °C)
Vapor pressure - 0,1 hPa (20 °C)
Storage temperature 15...25 °C
Assay (acidimetric) ≥99%
Melting range (lower value) ≥198%
Melting range (upper value) ≤204 °C
Identity (IR) passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR