Methyl ethyl ketone, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

8052.1000EA 56.7 EUR
8052.1000 8052.2500 8052.9025
Methyl ethyl ketone, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99 %
Acidity (as CH3COOH) max. 0.003 %
Boiling Range max. 1.5 °C
Density (g/ml) at 25°C 0.797 - 801.000
Recorded Boiling Point 79.6 °C
Residue after Evaporation max. 0.005 %
Water (H2O) max. 0.2 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR