Supelco® Organic Reference Standard, (−)-α-Pinene

Supplier: Merck

80599-1ML 80599-5ML
80599-1MLEA 56.9 EUR
80599-1ML 80599-5ML
Supelco® Organic Reference Standard, (−)-α-Pinene
Standards Organic Standards

  • GHS signal: GHS02, GHS07, GHS09


Caution: P210 - P233 - P273 - P301 + P310 - P303 + P361 + P353 - P331

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c72a77ed42616a094e4010e72bff0cb726a77c35&vid=f9c4e8dad72a913906603fadafb154f6ef893b5f&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) (1S,5S)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene; (1S,5S)-2-Pinene
Environmentally Preferable
M-Clarity quality level MQ100
Density 0,858 g/ml at 20 °C (lit.)
CAS No. 7785-26-4
MDL No. MFCD00064145
Molecular weight 136,23
Boiling point 156 - 158 °C (lit.)
Hazard class description H226 - H302 - H304 - H315 - H410
Flash point 31,3 °C
Shipping conditions STD
Storage conditions 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR