Xylene (mixture of isomers), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Dimethylbenzene

8080.1000EA 78.43 EUR
8080.1000 8080.2500 8080.9025 8080.5000
Xylene (mixture of isomers), BAKER ANALYZED® ACS, J.T.Baker®
Xylene (mixture of isomers)
Formula: C₆H₄(CH₃)₂
MW: 106.17 g/mol
Boiling Pt: 140
Melting Pt: –34 °C
Density: 0.86…0.88 g/cm³ (20 °C)
Flash Pt: 25
Storage Temperature: Ambient
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Exceeds ACS Specifications
Assay min. 98.5 %
Color (APHA) max. 10
Ethylbenzene max. 25 %
Residue after Evaporation max. 0.002 %
Sulfur Compounds (as S) max. 0.003 %
Substances Darkened by H2SO4 passes test
Water (H2O) max. 0.03 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR