Vanadium(III) fluoride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Vanadium trifluoride

81115.04 81115.09 81115.18
81115.04EA 160 EUR
81115.04 81115.09 81115.18
Vanadium(III) fluoride ≥98%
Vanadium (III) fluoride

Purity: 98% CAS No.: 10049-12-4 Molecular Formula: VF3 Formula Weight: 107.94 Physical Form: Powder Density: 3.363 Melting Point: ≈1406° Solubility: Practically insoluble in water, acetone, and glacial acetic acid Merck Reference: 14,9922 UN No.: 2923 MDL No.: MFCD00042553

Formula: F₃V
Melting Pt: ∼1406 °C
Density: 3,363
Storage Temperature: Ambient
MDL Number: MFCD00042553
CAS Number: 10049-12-4
EINECS: 233-169-6
UN: 2923
ADR: 8,II
Merck Index: 14,9922

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR