J.T.Baker® BAKERBOND Speedisk® Amino (NH2), SPE Columns
Supplier: Avantor
8165-03EA 0 EUR
8165-03
J.T.Baker® BAKERBOND Speedisk® Amino (NH2), SPE Columns
Chromatography Columns
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. Run times are up to nine times faster than conventional columns. The built-in column guard filter and higher analyte concentration may reduce or eliminate prefiltration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
General applications: Lipids (fatty acids, cholesterol).
These SPE columns can be used in ion exchange and normal phase mode.