Petroleum spirit, 60…100 °C, BAKER, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

8241.9025EA 0 EUR
8241.9025
Petroleum spirit, 60…100 °C, BAKER, Laboratory Reagent, J.T.Baker®
Petroleum spirit
Boiling Pt: 60…100 °C (1013 hPa)
Density: 0.67 g/cm³ (20 °C)
Flash Pt: -10
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
EINECS: 265-151-9
UN: 1268
ADR: 3 II

Order Now

Specification Test Results

Boiling Range 60 - 100 °C
Residue after Evaporation max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR