Mercury(II) sulphate 99,0 Precipitation titration, Puriss. p.a. ACS, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Mercury sulphate, Mercuric sulphate

83384-250G 83384-50G 83384-100G 83384-3KG
83384-250GEA 373 EUR
83384-250G 83384-50G 83384-100G 83384-3KG
Mercury(II) sulphate 99,0 Precipitation titration, Puriss. p.a. ACS, Fluka™
Mercury (II) sulphate
Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850 Dec.
Density: 7.56 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1 II
Merck Index: 13,05914

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Glowing After reducing with HCOOH(as SO₄) Max. 0.02 %
Calcium (Ca) Max. 10 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 30 ppm
Nitrate (NO₃) Max. 50 ppm
Mercury (Hg⁺) Max. 0.15 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR