Sodium benzoate 99.0-100.5% Ph. Eur.

Supplier: VWR Chemicals
Warning

83551.290EA 87.4 EUR
83551.290 83551.360 VWRC83551.461
Sodium benzoate 99.0-100.5% Ph. Eur.
Sodium benzoate
Formula: C₆H₅COONa
Melting Pt: 410…430 °C
Density: 1,44 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8
REACH: 01-2119460683-35
Merck Index: 13,08654

Order Now

Specification Test Results

Assay (calculated on dried substance) 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
Acidity or alkalinity Passes test
Halogenated compounds Passes test
Heavy metals (as Pb) Max. 10 ppm
Loss on drying (105°C) Max. 2 %
Statement of original manufacturer (*)
Benzene (*) Max. 2 ppm
Toluene (*) Max. 890 ppm
Other residual solvents (*) Unlikely by manuf.process

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR