Tetrahydrofuran, anhydrous (max. 0.003% H₂O) ≥99.9% stabilised

Supplier: VWR Chemicals
Danger


Stabilisation: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 250 ppm
Filtered through a 0.2 µm filter, packaged under nitrogen.

83678.230EA 162 EUR
83678.230 83678.290 83678.269
Tetrahydrofuran, anhydrous (max. 0.003% H₂O) ≥99.9% stabilised
Tetrahydrofuran

250 ml bottle with a septum cap featuring six separate re-sealable puncture points

Formula: C₄H₈O
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108,5 °C
Density: 0,888 g/cm³ (20 °C)
Flash Pt: –21,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99.9 %
Acidity Max. 0.0005 meq/g
Evaporation residue Max. 5 ppm
Water Max. 30 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR