Methanol, Wasser_0,002 99,9

Supplier: VWR Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filter 0,2 um_Nitrogen_filled

83679.230EA 78.3 EUR
83679.230 83679.290 83679.260
Methanol, Wasser_0,002 99,9
Methanol

250 ml bottle with a septum cap featuring six separate re-sealable puncture points

Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.9 %
Acidity Max. 0.00050 meq/g
Residue on evaporation Max. 0.0003 %
Water (K.F.) Max. 0.0020 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR