Toluene, Wasser_0,002 99,8

Supplier: VWR Chemicals
Danger

Synonyms: Methylbenzene, Phenylmethane, FM approval

Filter 0,2 um_Nitrogen_filled

83680.230EA 107 EUR
83680.230 83680.290
Toluene, Wasser_0,002 99,8
Toluene

250 ml bottle with a septum cap featuring six separate re-sealable puncture points

Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3 II
REACH: 01-2119471310-51
Merck Index: 13,09607
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44efa31292a7dce6d9e60bcbaec208eb42d34e32&vid=73ada9ad1e894808b6c193cabc4babcfaf320a25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated on anhydrous) Min. 99.8 %
Acidity Max. 0.0005 meq/g
Residue on evaporation Max. 0.0005 %
Water (K.F.) Max. 0.0020 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR