Ethanol ≥96% (v/v), TechniSolv®, pure

Supplier: VWR Chemicals
Danger

83804.410EA 177 EUR
83804.410 83804.360_T 83804.410_T 83804.360 83804.440 83804.550 83804.930
Ethanol ≥96% (v/v), TechniSolv®, pure
Ethanol

Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Formula: C₂H₅OH
Boiling Pt: 78,3 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0,7895 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II
REACH: 01-2119457610-43

Order Now

Specification Test Results

Assay Min. 95 %
Assay (V/V) Min. 95 %
Density (20/20) 0.805 - 0.811


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR