VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 22 Elements in Nitric Acid 5% for ICP
Supplier: VWR Chemicals
84790.180EA 420 EUR
84790.180
VWR® ARISTAR®, Multi-Element Quality Control Standard 100 mg/L, 22 Elements in Nitric Acid 5% for ICP
Standards Cationic Standards Multi-Element Standards
22 elements 100 mg/L: As, Be, Bi, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery information: Supplied with certificate of analysis.