System for parasitological examination of stool, ParasiTrap® Dual Eco
Supplier: Biosepar
8486DUALECO 8486DUALECO/50 8486DUALECO/10
8486DUALECOEA 0 EUR
8486DUALECO 8486DUALECO/50 8486DUALECO/10
System for parasitological examination of stool, ParasiTrap® Dual Eco
Clinical Diagnostic Supplies Clinical Reagents and Kits
ParasiTrap Dual Eco contains filled processing and transport tubes I (50 pieces Medium A Bailenger and 50 pieces Medium A ECO) and 100 pieces processing tubes II with integrated stainless steel filter system.
Accessories information: Separation medium B, staining solution medium C, Pasteur pipettes (disposable), spoons and cotton wool sticks. Additional screw caps for processing tube II.