Formic acid 99, HiPerSolv CHROMANORM® LC-MS

Supplier: VWR Chemicals
Danger

Synonyms: Aminic Acid, Methanoic acid

Nitrogen_filled

85048.001EA 91.8 EUR
85048.001 85048.010 85048.051
Formic acid 99, HiPerSolv CHROMANORM® LC-MS
Formic acid

LC-MS additives are selective chemicals that are commonly added to the mobile phase in LC-MS. The objective is usually to improve the signal quality, but they can also be used to suppress unwanted signals or selectively enhance the signal of specific compounds in a mixture


  • High quality and LC-MS application tested
  • Improved ionisation and resolution

Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 100…101 °C (1013 hPa)
Melting Pt: 8,2…8,4 °C
Density: 1.22 g/cm³ (25 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 (3) II
REACH: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on anhydrous substance) Min 99 %
Evaporation residue Max 0.0001 %
Ca (Calcium) (0.1 %) Max 0.2 ppm
K (Potassium) (0.1 %) Max 0.2 ppm
Mg (Magnesium) (0.1 %) Max 0.2 ppm
Na (Sodium) (0.1 %) Max 0.5 ppm
Transmittance (225 nm)(0.1 %) Min 20 %
Transmittance (235 nm)(0.1 %) Min 50 %
Transmittance (240 nm)(0.1 %) Min 80 %
Transmittance (250 nm)(0.1 %) Min 95 %
Transmittance (260 nm)(0.1 %) Min 99 %
Suitability for LC-MS (0.1 %) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR