Ammonium carbonate in aqueous solution USP Test Solution (TS)

Supplier: VWR Chemicals

Synonyms: Salt of hartshorn

85328.180EA 82.3 EUR
85328.180 85328.260
Ammonium carbonate in aqueous solution USP Test Solution (TS)
Ammonium carbonate

Ready to use test solution (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Manufactured with controlled processes
  • Independent, traceable and certified
  • Certificates of analysis and safety data sheets available

Formula: (NH₄)₂CO₃
MW: 96.09 g/mol
Melting Pt: 58
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 0
Merck Index: 13,00508

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR