Potassium acetate aqueous solution USP Test Solution (TS) 34

Supplier: VWR Chemicals

Synonyms: Acetic acid potassium salt

85364.260EA 119 EUR
85364.260
Potassium acetate aqueous solution USP Test Solution (TS) 34
Potassium acetate

Ready to use test solutions (TS).


  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available

Formula: CH₃COOK
MW: 98.14 g/mol
Melting Pt: 304
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 0
Merck Index: 13,07687
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR