N-Methyl-2-pyrrolidone (NMP) 99,8, PESTINORM® GC_HS

Supplier: VWR Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

85398.290EA 308 EUR
85398.290
N-Methyl-2-pyrrolidone (NMP) 99,8, PESTINORM® GC_HS
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
REACH: 01-2119472430-46
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Limited Time Sale

Select items below are currently on sale

Order Now

Specification Test Results

Purity (GC) min. 99.8%
Identity (IR) Complying
Water (KF) max. 0.05%w/w
Color (APHA Pt-Co) max. 10
GC-HS (Each class 1 solvent) max. 1µg/g
GC-HS (Each class 2 solvent) max. 10µg/g
GC-HS (Each class 3 solvent) max. 50µg/g
GC-HS (Each unknown, MIBK-DMSO) max. 10µg/g
Suitable as solvent for the analysis of residual solvents ofICH classes 1, 2 and 3 according to Ph Eur and USP. Passes test
Each unknown measured against butyl methyl ketone up to DMSOis ≤ 10 µg/g.
Formic acid and acetic acid are not detected.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR