Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)

Supplier: VWR Chemicals

85825.360EA 55.2 EUR
85825.360 85825.440 85825.550
Ethanol eurodenatured ≥70%, TechniSolv® (1:1:1)
Ethanol Eurodenatured
Formula: H₃CCH₂OH
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Assay (V/V) min. 68.4%
Denatonium benzoate (ppm) 7.9 - 9.2 ppm
Methyl ethyl ketone (V/V) 0.68 - 0.80 %
2-Propanol (V/V) 0.68 - 0.80%

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental certification EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR