Triethylamine 99, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: (Diethylamino)ethane, TEA

85908.180EA 55.7 EUR
85908.180 85908.260 85908.290 85908.320 85908.460
Triethylamine 99, GPR RECTAPUR®
Triethylamine
Formula: (C₂H₅)₃N
MW: 101.19 g/mol
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –115 °C
Density: 0.729 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00009051
CAS Number: 121-44-8
EINECS: 204-469-4
UN: 1296
ADR: 3 (8)
Merck Index: 13,09740

Order Now

Specification Test Results

Assay ≥ 99%
IR Spectrum Passes test
Refractive index (20°C) 1,399 - 1,401
Water ≤ 0,2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR