N,N-Dimethyl-p-toluidine ≥98.0% (by GC), GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: N,N,4-Trimethylaniline, N,N-Dimethyl-4-methylaniline, 1-(Dimethylamino)-4-methylbenzol, 4,N,N-Trimethylaniline, 4-(Dimethylamino)toluene, 4-Dimethylaminotoluene

85986.230EA 84.9 EUR
85986.230 85986.290 85986.460
N,N-Dimethyl-p-toluidine ≥98.0% (by GC), GPR RECTAPUR®
N,N-Dimethyl-p-toluidine
Formula: C₉H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD00008316
CAS Number: 99-97-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear pale yellow to amber liquid
Assay (GC) min. 98,0 %
Water max. 0,5 %
Density (d20°C/4°C) 0,935 – 0,937
Identity (IR) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR