1-Hexanol ≥98.0% (by GC), GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

85998.180EA 24.1 EUR
85998.180 85998.290 85998.320
1-Hexanol ≥98.0% (by GC), GPR RECTAPUR®
1-Hexanol
Formula: H₃C(CH₂)₄CH₂OH
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00002982
CAS Number: 111-27-3
EINECS: 203-852-3
UN: 2282
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0793e56caf8b4e459c31d9f36b2eec3d50a4e4&vid=5117270d1d57dd4b9cca9ee3d787375106cc1216&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Assay (GC) min. 98.0 %
Density (d20°C/ 4°C) 0.817 - 0.821
Identity (IR) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR