Hanus solution (Iodine bromine) 0.1 mol/l (0.2 N), AVS TITRINORM Iodine_value

Supplier: VWR Chemicals
Danger

Synonyms: Iodine solution acc. to Hanus

86182.290EA 114 EUR
86182.290
Hanus solution (Iodine bromine) 0.1 mol/l (0.2 N), AVS TITRINORM Iodine_value
Hanus solution (Iodine bromine)
Formula: IBr
Storage Temperature: Ambient
MDL Number: MFCD00011353
UN: 2789
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR