Copper di-ammonium EDTA 0.1 mol/l, AVS TITRINORM

Supplier: VWR Chemicals

Synonyms: Ethylenediaminetetraacetic acid diammonium copper salt

86186.260EA 116 EUR
86186.260
Copper di-ammonium EDTA 0.1 mol/l, AVS TITRINORM
Copper di-ammonium EDTA
Formula: C₁₀H₂₀CuN₄O₈
MW: 387.83 g/mol
MDL Number: MFCD01863036
CAS Number: 67989-88-2

Order Now

Specification Test Results

di-ammonium copper EDTA 0.1 mol/l
Appearance clear
Colour Blue
Density (20 °C) 1,020 – 1,030 g/ml
Not suitable for samples containing silver.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR