Potassium hydroxide solution_SC 0.1 mol/l (0.1 N) 2-propanol acc. to DIN 51558 part 1, AVS TITRINORM

Supplier: VWR Chemicals

Synonyms: Potash caustic, Caustic potash

86190.290EA 79.8 EUR
86190.290
Potassium hydroxide solution_SC 0.1 mol/l (0.1 N) 2-propanol acc. to DIN 51558 part 1, AVS TITRINORM
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
MDL Number: MFCD00003553
CAS Number: 1310-58-3
Merck Index: 12,07806

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR