VWR®, Gallium standard solution, 1,000 mg/L Ga in dilute nitric acid AVS TITRINORM standard for AAS
Supplier: VWR Chemicals
86678.180EA 94.6 EUR
86678.180 86678.260
VWR®, Gallium standard solution, 1,000 mg/L Ga in dilute nitric acid AVS TITRINORM standard for AAS
Standards Cationic Standards Gallium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%
Order Now
SPECIFICATIONS
Environmentally Preferable