VWR® AVS TITRINORM, Iridium Standard Solution, 1,000 mg/L Ir in 10% Hydrochloric Acid AVS TITRINORM Standard for AAS
Supplier: VWR Chemicals
86685.180EA 177.8 EUR
86685.180 86685.260
VWR® AVS TITRINORM, Iridium Standard Solution, 1,000 mg/L Ir in 10% Hydrochloric Acid AVS TITRINORM Standard for AAS
Standards Cationic Standards Iridium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%