VWR® AVS TITRINORM, Thallium Standard Solution, 1000 mg/L Tl in dil. Nitric Acid, Standard for AAS
Supplier: VWR Chemicals
86718.180EA 34.3 EUR
86718.180 86718.260
VWR® AVS TITRINORM, Thallium Standard Solution, 1000 mg/L Tl in dil. Nitric Acid, Standard for AAS
Standards Cationic Standards Thallium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%