Tetrabutylammonium hydrogen sulphate 99,0 Ionpair, Supelco®

Supplier: Merck

Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate

86853-10G-F 86853-50G-F 86853-10X1G-F
86853-10G-FEA 301 EUR
86853-10G-F 86853-50G-F 86853-10X1G-F
Tetrabutylammonium hydrogen sulphate 99,0 Ionpair, Supelco®
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
MDL Number: MFCD00011637
CAS Number: 32503-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ac945220fa3730d357572c05c3735d620538336&vid=4ac945220fa3730d357572c05c3735d620538336&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR