4-(Dimethylamino)pyridine ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine

87080.180EA 111 EUR
87080.180 87080.130
4-(Dimethylamino)pyridine ≥99%, GPR RECTAPUR®
4-(Dimethylamino)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 195 °C (1013 hPa)
Melting Pt: 109…111 °C
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Assay Min. 99 %
Melting point 111 --> 114 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR