Isonicotinic acid, GPR RECTAPUR®

Supplier: VWR Chemicals
Warning

87093.230EA 83.6 EUR
87093.230
Isonicotinic acid, GPR RECTAPUR®
Isonicotinic acid
Formula: C₆H₅NO₂
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 315…319 °C
Storage Temperature: Ambient
MDL Number: MFCD00006429
CAS Number: 55-22-1
EINECS: 200-228-2
Merck Index: 13,05206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR