Anisole ≥99%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Methyl phenyl ether, Methoxybenzene

87106.180EA 33.5 EUR
87106.180 87106.260 87106.290 87106.320 87106.460
Anisole ≥99%, GPR RECTAPUR®
Anisole
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 154
Melting Pt: -37
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
UN: 2222
ADR: 3 III
Merck Index: 13,00672

Order Now

Specification Test Results

Assay ≥ 99 %
Appearance Clear colourless liquid
Odour Characteristic
Density (20/4) 0,992 - 0,994


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR