Copper (II) ethylenediamine 21% in aqueous solution Reag. Ph. Eur. 3008700

Supplier: VWR Chemicals
Danger

Synonyms: Bis(ethylenediamine)copper dihydroxide, Cupriethylendiamine, Copper(II)-ethylenediame complex

87712.290EA 691 EUR
87712.290
Copper (II) ethylenediamine 21% in aqueous solution Reag. Ph. Eur. 3008700
Copper (II) ethylenediamine

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MDL Number: MFCD00274628
CAS Number: 14552-35-3
EINECS: 238-597-7
UN: 1761
ADR: 8 (6.1),II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR