Acetic acid 30% Reag. Ph. Eur. 1000401

Supplier: VWR Chemicals
Danger

87758.290EA 98.3 EUR
87758.290
Acetic acid 30% Reag. Ph. Eur. 1000401
Acetic acid

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: H₃CCOOH
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2790
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR