Barium chloride 0.25 mol/l (0.5 N) in aqueous solution Reag. Ph. Eur. 1009301

Supplier: VWR Chemicals
Warning

87783.290EA 369 EUR
87783.290
Barium chloride 0.25 mol/l (0.5 N) in aqueous solution Reag. Ph. Eur. 1009301
Barium chloride

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: BaCl₂
MW: 208.24 g/mol
Boiling Pt: 1560
Melting Pt: 963
Density: 1.034 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1 III
Merck Index: 12,00998

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR