Hydrochloric acid brominated Reag. Ph. Eur. 1043507

Supplier: VWR Chemicals
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

87845.290EA 168 EUR
87845.290
Hydrochloric acid brominated Reag. Ph. Eur. 1043507
Hydrochloric acid

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: HCl
MW: 36.46 g/mol
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1760
ADR: 8 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR