Eriochrome Black T 1% in sodium chloride Reag. Ph. Eur. 1056801

Supplier: VWR Chemicals
Warning

87873.180EA 89.2 EUR
87873.180
Eriochrome Black T 1% in sodium chloride Reag. Ph. Eur. 1056801
Eriochrome Black T

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: C₂₀H₁₂N₃NaO₇S
Storage Temperature: Ambient
MDL Number: MFCD00003935
CAS Number: 1787-61-7
EINECS: 217-250-3
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR