Pararosaniline solution_SC Reag. Ph. Eur. 1062201, decolourised

Supplier: VWR Chemicals
Danger

Synonyms: Tris(4-aminophenyl)methanol, Paramagenta, p-Rosaniline, Parafuchsin, Pararosaniline base

87885.180EA 108 EUR
87885.180
Pararosaniline solution_SC Reag. Ph. Eur. 1062201, decolourised
Pararosaniline

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: C₁₉H₁₉N₃O
MW: 305.38 g/mol
Boiling Pt: ~100 °C (1013 hPa)
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036222
CAS Number: 467-62-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR