Potassium hexacyanoferrate(II) trihydrate 5,3% (w/v) in aqueous solution Reag. Ph. Eur. 1069801

Supplier: VWR Chemicals

Synonyms: Potassiumhexacyanoferrate(II) trihydrate, Tetrapotassium hexacyanoferrate trihydrate, Potassium ferrocyanide trihydrate

87902.180EA 121 EUR
87902.180
Potassium hexacyanoferrate(II) trihydrate 5,3% (w/v) in aqueous solution Reag. Ph. Eur. 1069801
Potassium hexacyanoferrate (II) trihydrate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: K₄Fe(CN)₆·3H₂O
MW: 422.39 g/mol
Melting Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00167023
CAS Number: 14459-95-1
EINECS: 237-722-2
Merck Index: 12,07793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR