Potassium hydroxide 3% in ethanol 95% Reag. Ph. Eur. 1070303

Supplier: VWR Chemicals

87906.180EA 94.3 EUR
87906.180
Potassium hydroxide 3% in ethanol 95% Reag. Ph. Eur. 1070303
Potassium hydroxide

Ordering information: Normally manufactured to order. Please check with customer services.

Formula: KOH
MDL Number: MFCD00003553
CAS Number: 1310-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR