Sodium hydroxide 20% w/w aqueous solution Reag. Ph. Eur. 1081401

Supplier: VWR Chemicals
Danger

Synonyms: Caustic soda

87933.290EA 85.2 EUR
87933.290
Sodium hydroxide 20% w/w aqueous solution Reag. Ph. Eur. 1081401
Sodium hydroxide

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 323
Density: ∼1.22 g/ml
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Limited Time Sale

Select items below are currently on sale

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR