Sulphuric acid concentrate for dilution to 2.0 N aqueous solution, Acculute™ Standard Volumetric Solution

Supplier: Thermo Fisher Scientific
Danger

88568.LQ
88568.LQEA 87.3 EUR
88568.LQ
Sulphuric acid concentrate for dilution to 2.0 N aqueous solution, Acculute™ Standard Volumetric Solution
Sulphuric acid

CAS No.: 7664-93-9 Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Appearance: Colourless to pale yellow Odour: Acrid UN No.: 2796 MDL No.: MFCD00064589

Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR