Silver trifluoromethanesulphonate ≥98%

Supplier: Thermo Fisher Scientific
Danger

88722.04 88722.09 88722.18
88722.04EA 44.1 EUR
88722.04 88722.09 88722.18
Silver trifluoromethanesulphonate ≥98%
Silver trifluoromethanesulphonate

Purity: 98% CAS No.: 2923-28-6 Molecular Formula: CAgF3O3S Formula Weight: 256.93 Physical Form: Crystalline Sensitivity: Light Sensitive Appearance: White to grey Odour: Odourless Melting Point: 356° UN No.: 3260 MDL No.: MFCD00013226

Formula: CAgF₃O₃S
Melting Pt: 356 °C
Storage Temperature: Ambient
MDL Number: MFCD00013226
CAS Number: 2923-28-6
EINECS: 220-882-2
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR