Amberlyst® 15 (H) ion exchange resin

Supplier: Thermo Fisher Scientific

89079.18 89079.30 89079.A1
89079.18EA 36.1 EUR
89079.18 89079.30 89079.A1
Amberlyst® 15 (H) ion exchange resin
Amberlyst® 15 (H)

CAS No.: 39389-20-3 Molecular Formula: Styrene-DVB; Strongly acidic macroreticular resin Physical Form: Beads Appearance: Pale brown Odour: Odourless MDL No.: MFCD00145841

Formula: (C₁₀H₁₀·C₈H₈O₃S)x
Storage Temperature: Ambient
MDL Number: MFCD00145841
CAS Number: 39389-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR