Sodium perchlorate monohydrate ≥99.0% for HPLC, Fluka™

Supplier: Honeywell Chemicals
Danger

89152-250G 89152-50G
89152-250GEA 254 EUR
89152-250G 89152-50G
Sodium perchlorate monohydrate ≥99.0% for HPLC, Fluka™
Sodium perchlorate monohydrate
Formula: NaClO₄·H₂O
Boiling Pt: Decomposes on heating
Melting Pt: 469 °C (anhydr.)
Density: 2,02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149164
CAS Number: 7791-07-3
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 12,08798

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Solution 5% in H₂O complying
pH (5 %, 25 °C) 5.0 - 8.0
Absorbance at 210 nm Max. 0.1
Absorbance at 220 nm Max. 0.05
Absorbance at 230 nm Max. 0.03
Absorbance at 240 nm Max. 0.02
Absorbance 500 nm Max. 0.02


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR