Titanium(IV) isobutoxide

Supplier: Thermo Fisher Scientific
Danger

89455.18 89455.30 89455.A1
89455.18EA 46.9 EUR
89455.18 89455.30 89455.A1
Titanium(IV) isobutoxide
Titanium(IV) isobutoxide

CAS No.: 681-98-1 Molecular Formula: C16H36O4Ti Formula Weight: 340.34 Flash Point: 77°(171°F) Physical Form: Melt Density: 1.02 Refractive Index: 1.4950 Sensitivity: Moisture Sensitive Appearance: Yellow Boiling Point: 296° MDL No.: MFCD00015045

Formula: C₁₆H₃₆O₄Ti
Boiling Pt: 296 °C
Density: 1,02
Flash Pt: 77 °C (171 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015045
CAS Number: 681-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR